Anti-PDE4C Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
WB:1:500-1:2000, ELISA:1:5000
Type:
Antigen: PDE4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77720-245
- 77705-630
- Antigen Symbol:
- PDE4C
- PDE4C
- Antigen Synonyms:
- Pde4c|5''-cyclic phosphodiesterase 4C|cAMP specific 3' 5' cyclic phosphodiesterase 4C|PDE21|Phosphodiesterase 4C|PDE 21|PDE4C_HUMAN|Phosphodiesterase 4C, cAMP specific (phosphodiesterase E1 dunce homolog, Drosophila)|Phosphodiesterase E1 dunce homolog|Phosphodiesterase E1 dunce homolog Drosophila|PDE 4C|MGC126222|DPDE 1|DPDE1|Phosphodiesterase 4C cAMP specific|cAMP-specific 3''|Phosphodiesterase type 4C
- Pde4c|5''-cyclic phosphodiesterase 4C|cAMP specific 3' 5' cyclic phosphodiesterase 4C|PDE21|Phosphodiesterase 4C|PDE 21|PDE4C_HUMAN|Phosphodiesterase 4C, cAMP specific (phosphodiesterase E1 dunce homolog, Drosophila)|Phosphodiesterase E1 dunce homolog|Phosphodiesterase E1 dunce homolog Drosophila|PDE 4C|MGC126222|DPDE 1|DPDE1|Phosphodiesterase 4C cAMP specific|cAMP-specific 3''|Phosphodiesterase type 4C
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q08493
- Q08493
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthesized peptide derived from the Internal region of Human PDE4C
- Synthesized peptide derived from the Internal region of Human PDE4C
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together