Anti-NCR2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, IHC:1:50-1:200
Type:
Antigen: NCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-212
- 77720-213
- Antigen Symbol:
- NCR2
- NCR2
- Antigen Synonyms:
- LY95|Activating NK receptor NKp44|NK cell activating receptor|CD336|NK p44|Lymphocyte antigen 95 homolog|Natural killer cell p44 related protein|NK cell-activating receptor|Lymphocyte antigen 95 homolog (activating NK receptor, NK p44)|Lymphocyte antigen 95 (activating NK receptor, NK p44)|NK-p44|NK cell activating receptor (NKp44)|Lymphocyte antigen 95, mouse, homolog of|NKp44|NCR2|CD336 antigen|Natural killer cell p44-related protein|Natural cytotoxicity triggering receptor 2|dJ149M18.1|NCTR2_HUMAN
- LY95|Activating NK receptor NKp44|NK cell activating receptor|CD336|NK p44|Lymphocyte antigen 95 homolog|Natural killer cell p44 related protein|NK cell-activating receptor|Lymphocyte antigen 95 homolog (activating NK receptor, NK p44)|Lymphocyte antigen 95 (activating NK receptor, NK p44)|NK-p44|NK cell activating receptor (NKp44)|Lymphocyte antigen 95, mouse, homolog of|NKp44|NCR2|CD336 antigen|Natural killer cell p44-related protein|Natural cytotoxicity triggering receptor 2|dJ149M18.1|NCTR2_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95944
- O95944
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human NCR2
- Synthetic peptide of Human NCR2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together