Order Entry
United States
ContactUsLinkComponent
Anti-BIRC2 Rabbit Polyclonal Antibody
Anti-BIRC2 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-BIRC2 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77720-170
  • 77720-171
  • Antigen Symbol:
  • BIRC2
  • BIRC2
  • Antigen Synonyms:
  • TNFR2-TRAF-signaling complex protein 2|HIAP2|BIRC2_HUMAN|IAP homolog B|TNFR2 TRAF signaling complex protein 2|Inhibitor of apoptosis protein 2|NFR2 TRAF signalling complex protein|HIAP 2|C-IAP1|RNF48|cellular inhibitor of apoptosis protein 1|cIAP 1|API1|IAP 2|HIAP-2|RING finger protein 48|Baculoviral IAP repeat containing protein 2|C IAP1|Baculoviral IAP repeat-containing protein 2|cIAP1|IAP-2|BIRC2|Baculoviral IAP repeat containing 2|API 1|Cellular inhibitor of apoptosis 1|MIHB|RNF 48|Apoptosis inhibitor 1|IAP2|BIRC 2
  • TNFR2-TRAF-signaling complex protein 2|HIAP2|BIRC2_HUMAN|IAP homolog B|TNFR2 TRAF signaling complex protein 2|Inhibitor of apoptosis protein 2|NFR2 TRAF signalling complex protein|HIAP 2|C-IAP1|RNF48|cellular inhibitor of apoptosis protein 1|cIAP 1|API1|IAP 2|HIAP-2|RING finger protein 48|Baculoviral IAP repeat containing protein 2|C IAP1|Baculoviral IAP repeat-containing protein 2|cIAP1|IAP-2|BIRC2|Baculoviral IAP repeat containing 2|API 1|Cellular inhibitor of apoptosis 1|MIHB|RNF 48|Apoptosis inhibitor 1|IAP2|BIRC 2
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q13490
  • Q13490
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Fusion protein of Human BIRC2
  • Fusion protein of Human BIRC2
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together