Anti-UHRF2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: UHRF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77720-160
- 77720-159
- Antigen Symbol:
- UHRF2
- UHRF2
- Antigen Synonyms:
- Nuclear zinc finger protein Np97|Np95 like RING finger protein|E3 ubiquitin-protein ligase UHRF2|Np95-like ring finger protein|Np95/ICBP90 like RING finger protein|Ubiquitin-like-containing PHD and RING finger domains protein 2|DKFZp434B0920|UHRF2_HUMAN|RP11-472F14.2|RING finger protein 107|MGC33463|Np95/ICBP90-like RING finger protein|Ubiquitin like containing PHD and RING finger domains protein 2|RNF 107|DKFZp686G0837|Uhrf2|Nuclear protein 97|Ubiquitin-like PHD and RING finger domain-containing protein 2|URF 2
- Nuclear zinc finger protein Np97|Np95 like RING finger protein|E3 ubiquitin-protein ligase UHRF2|Np95-like ring finger protein|Np95/ICBP90 like RING finger protein|Ubiquitin-like-containing PHD and RING finger domains protein 2|DKFZp434B0920|UHRF2_HUMAN|RP11-472F14.2|RING finger protein 107|MGC33463|Np95/ICBP90-like RING finger protein|Ubiquitin like containing PHD and RING finger domains protein 2|RNF 107|DKFZp686G0837|Uhrf2|Nuclear protein 97|Ubiquitin-like PHD and RING finger domain-containing protein 2|URF 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96PU4
- Q96PU4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human UHRF2
- Synthetic peptide of Human UHRF2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together