Anti-BIRC7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000
Type:
Antigen: BIRC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-141
- 77720-140
- Antigen Symbol:
- BIRC7
- BIRC7
- Antigen Synonyms:
- MLIAP|Livin|Baculoviral IAP repeat-containing protein 7|ML IAP|Kidney inhibitor of apoptosis protein|Livin inhibitor of apotosis|Melanoma inhibitor of apoptosis protein|RING finger protein 50|RNF50|birc7|Baculoviral IAP repeat containing 7|BIRC 7|ML-IAP|BIRC7_HUMAN|RNF 50|KIAP|Baculoviral IAP repeat containing protein 7
- MLIAP|Livin|Baculoviral IAP repeat-containing protein 7|ML IAP|Kidney inhibitor of apoptosis protein|Livin inhibitor of apotosis|Melanoma inhibitor of apoptosis protein|RING finger protein 50|RNF50|birc7|Baculoviral IAP repeat containing 7|BIRC 7|ML-IAP|BIRC7_HUMAN|RNF 50|KIAP|Baculoviral IAP repeat containing protein 7
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96CA5
- Q96CA5
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Full length fusion protein
- Full length fusion protein
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together