Order Entry
United States
ContactUsLinkComponent
Anti-RPS6KA6 Rabbit Polyclonal Antibody
Anti-RPS6KA6 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-RPS6KA6 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77720-109
  • 77720-108
  • Antigen Symbol:
  • RPS6KA6
  • RPS6KA6
  • Antigen Synonyms:
  • S6K-alpha-6|Ribosomal protein S6 kinase alpha-6|Ribosomal protein S6 kinase alpha 6|p90RSK6|p90 RSK 6|pp90RSK4|RSK-4|90 kDa ribosomal protein S6 kinase 6|Ribosomal protein S6 kinase 90kDa polypeptide 6|KS6A6_HUMAN|p90-RSK 6|RSK 4|rps6ka6|Ribosomal S6 kinase 4
  • S6K-alpha-6|Ribosomal protein S6 kinase alpha-6|Ribosomal protein S6 kinase alpha 6|p90RSK6|p90 RSK 6|pp90RSK4|RSK-4|90 kDa ribosomal protein S6 kinase 6|Ribosomal protein S6 kinase 90kDa polypeptide 6|KS6A6_HUMAN|p90-RSK 6|RSK 4|rps6ka6|Ribosomal S6 kinase 4
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UK32
  • Q9UK32
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide of Human RPS6KA6
  • Synthetic peptide of Human RPS6KA6
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together