Anti-CAGE1 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, IHC:1:25-1:100
Type:
Antigen: CAGE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77720-105
- 77720-104
- Antigen Symbol:
- CAGE1
- CAGE1
- Antigen Synonyms:
- Cancer/testis antigen 3|CT3|CAGE-1|CAGE1|CTAG3Cancer-associated gene 1 protein
- Cancer/testis antigen 3|CT3|CAGE-1|CAGE1|CTAG3Cancer-associated gene 1 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TC20
- Q8TC20
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CAGE1
- Synthetic peptide of Human CAGE1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together