Anti-FAM3B Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: FAM3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77720-099
- 77720-098
- Antigen Symbol:
- FAM3B
- FAM3B
- Antigen Synonyms:
- FAM3B_HUMAN|Protein FAM3B|2-21|Cytokine like protein 2 21|C21orf76|Fam3b|C21orf11|PANDER|ORF9|Cytokine-like protein 2-21|Pancreatic-derived factor|Family with sequence similarity 3 member B|PRED44
- FAM3B_HUMAN|Protein FAM3B|2-21|Cytokine like protein 2 21|C21orf76|Fam3b|C21orf11|PANDER|ORF9|Cytokine-like protein 2-21|Pancreatic-derived factor|Family with sequence similarity 3 member B|PRED44
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P58499
- P58499
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human FAM3B
- Fusion protein of Human FAM3B
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together