Anti-TIMP1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, IHC:1:15-1:50
Type:
Antigen: TIMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-078
- 77720-079
- Antigen Symbol:
- TIMP1
- TIMP1
- Antigen Synonyms:
- TIMP 1|Human Collagenase Inhibitor|TIMP metallopeptidase inhibitor 1|Metalloproteinase inhibitor 1|Tissue Inhibitor of Metalloproteinase 1|TIMP1_HUMAN|Erythroid Potentiating Activity|Fibroblast collagenase inhibitor|EPO|FLJ90373|Ttissue inhibitor of metalloproteinase 1 erythroid potentiating activity collagenase inhibitor|Timp1|Clgi|Tissue inhibitor of metalloproteinases|Metalloproteinase inhibitor 1 precursor|TIMP|HCI|OTTHUMP00000023214|Human|Collagenase inhibitor|EPA|Erythroid-potentiating activity|TIMP-1|Tissue inhibitor of metalloproteinases 1|TIMP1 protein
- TIMP 1|Human Collagenase Inhibitor|TIMP metallopeptidase inhibitor 1|Metalloproteinase inhibitor 1|Tissue Inhibitor of Metalloproteinase 1|TIMP1_HUMAN|Erythroid Potentiating Activity|Fibroblast collagenase inhibitor|EPO|FLJ90373|Ttissue inhibitor of metalloproteinase 1 erythroid potentiating activity collagenase inhibitor|Timp1|Clgi|Tissue inhibitor of metalloproteinases|Metalloproteinase inhibitor 1 precursor|TIMP|HCI|OTTHUMP00000023214|Human|Collagenase inhibitor|EPA|Erythroid-potentiating activity|TIMP-1|Tissue inhibitor of metalloproteinases 1|TIMP1 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01033
- P01033
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TIMP1
- Fusion protein of Human TIMP1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together