Anti-RAB8B Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:200-1:1000, IHC:1:30-1:150
Type:
Antigen: RAB8B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77720-027
- 77720-026
- Antigen Symbol:
- RAB8B
- RAB8B
- Antigen Synonyms:
- RAB44|RAB 8b protein|GTPase Rab8b|RAB8B, member RAS oncogene family|Ras-related protein Rab-8B|5930437D16|FLJ38125|D330025I23Rik|RAB8B_HUMAN|Rab8b
- RAB44|RAB 8b protein|GTPase Rab8b|RAB8B, member RAS oncogene family|Ras-related protein Rab-8B|5930437D16|FLJ38125|D330025I23Rik|RAB8B_HUMAN|Rab8b
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92930
- Q92930
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human RAB8B
- Synthetic peptide of Human RAB8B
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together