Anti-PRKAR1B Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:10000, WB:1:1000-1:5000, IHC:1:50-1:200
Type:
Antigen: PRKAR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77719-986
- 77719-987
- Antigen Symbol:
- PRKAR1B
- PRKAR1B
- Antigen Synonyms:
- cAMP dependent protein kinase type I beta regulatory subunit|cAMP-dependent protein kinase type I-beta regulatory subunit|Prkar1b|Protein kinase cAMP dependent regulatory type I beta|KAP1_HUMAN|RIbeta|PRKAR1|PRKAR1B protein|PRKAR 1|PKARI beta|PRKAR 1B
- cAMP dependent protein kinase type I beta regulatory subunit|cAMP-dependent protein kinase type I-beta regulatory subunit|Prkar1b|Protein kinase cAMP dependent regulatory type I beta|KAP1_HUMAN|RIbeta|PRKAR1|PRKAR1B protein|PRKAR 1|PKARI beta|PRKAR 1B
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31321
- P31321
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human PRKAR1B
- Synthetic peptide of Human PRKAR1B
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together