Anti-MAGEE1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:200-1:1000, IHC:1:50-1:200
Type:
Antigen: MAGEE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-985
- 77719-984
- Antigen Symbol:
- MAGEE1
- MAGEE1
- Antigen Synonyms:
- HCA1|MAGE-E1 antigen|DAMAGE|KIAA1587Melanoma-associated antigen E1|Hepatocellular carcinoma-associated protein 1|MAGEE1|Alpha-dystrobrevin-associated MAGE Protein
- HCA1|MAGE-E1 antigen|DAMAGE|KIAA1587Melanoma-associated antigen E1|Hepatocellular carcinoma-associated protein 1|MAGEE1|Alpha-dystrobrevin-associated MAGE Protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HCI5
- Q9HCI5
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human MAGEE1
- Synthetic peptide of Human MAGEE1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together