Anti-SLC20A1 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: SLC20A1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77719-890
- 77719-891
- Antigen Symbol:
- SLC20A1
- SLC20A1
- Antigen Synonyms:
- Leukemia virus receptor 1 homolog|FLJ41426|GLVR 1|Slc20a1|Gibbon ape leukemia virus receptor 1|Solute carrier family 20 (phosphate transporter) member 1|PIT 1|Sodium dependent phosphate transporter 1|Sodium-dependent phosphate transporter 1|Solute carrier family 20 member 1|S20A1_HUMAN|GLVR1|PiT-1|GLVR-1|Phosphate transporter 1|PIT1|DKFZp686J2397
- Leukemia virus receptor 1 homolog|FLJ41426|GLVR 1|Slc20a1|Gibbon ape leukemia virus receptor 1|Solute carrier family 20 (phosphate transporter) member 1|PIT 1|Sodium dependent phosphate transporter 1|Sodium-dependent phosphate transporter 1|Solute carrier family 20 member 1|S20A1_HUMAN|GLVR1|PiT-1|GLVR-1|Phosphate transporter 1|PIT1|DKFZp686J2397
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WUM9
- Q8WUM9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sodium-dependent phosphate transporter 1 protein (263-458AA)
- Recombinant Human Sodium-dependent phosphate transporter 1 protein (263-458AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together