Anti-SLC22A17 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:100-1:500, IF:1:50-1:500
Type:
Antigen: SLC22A17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-880
- 77690-187
- Antigen Symbol:
- SLC22A17
- SLC22A17
- Antigen Synonyms:
- Brain-type organic cation transporter|NGALR3|Solute carrier family 22 organic cation transporter member 17|OTTHUMP00000082690|BOIT|NgalR|Solute carrier family 22 member 17|BOCT|OTTHUMP00000082691|hBOIT|Slc22a17|Neutrophil gelatinase associated lipocalin receptor|Potent brain type organic ion transporter|S22AH_HUMAN|NGALR2|Neutrophil gelatinase-associated lipocalin receptor|Brain type organic cation transporter|NGAL receptor|Lipocalin-2 receptor|24p3R|24p3 receptor
- Brain-type organic cation transporter|NGALR3|Solute carrier family 22 organic cation transporter member 17|OTTHUMP00000082690|BOIT|NgalR|Solute carrier family 22 member 17|BOCT|OTTHUMP00000082691|hBOIT|Slc22a17|Neutrophil gelatinase associated lipocalin receptor|Potent brain type organic ion transporter|S22AH_HUMAN|NGALR2|Neutrophil gelatinase-associated lipocalin receptor|Brain type organic cation transporter|NGAL receptor|Lipocalin-2 receptor|24p3R|24p3 receptor
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WUG5
- Q8WUG5
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Solute carrier family 22 member 17 protein (1-99AA)
- Recombinant Human Solute carrier family 22 member 17 protein (1-99AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together