Anti-DENND1A Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DENND1A
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-845
- 77719-846
- Antigen Symbol:
- DENND1A
- DENND1A
- Antigen Synonyms:
- RP11 230L22.3|FAM31A|DENN/MADD domain containing 1A|dennd1a|Connecdenn|FLJ21129|OTTHUMP00000022090|Protein FAM31A|KIAA1608|FLJ38464|DENN domain-containing protein 1A|Connecdenn 1|DEN1A_HUMAN
- RP11 230L22.3|FAM31A|DENN/MADD domain containing 1A|dennd1a|Connecdenn|FLJ21129|OTTHUMP00000022090|Protein FAM31A|KIAA1608|FLJ38464|DENN domain-containing protein 1A|Connecdenn 1|DEN1A_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TEH3
- Q8TEH3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DENN domain-containing protein 1A protein (1-559AA)
- Recombinant Human DENN domain-containing protein 1A protein (1-559AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together