Anti-EPS8L3 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: EPS8L3
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77719-836
- 77719-835
- Antigen Symbol:
- EPS8L3
- EPS8L3
- Antigen Synonyms:
- EPS8-like protein 3|EPS8L3|EPS8-related protein 3|Epidermal growth factor receptor pathway substrate 8-related protein 3|EPS8R3Epidermal growth factor receptor kinase substrate 8-like protein 3
- EPS8-like protein 3|EPS8L3|EPS8-related protein 3|Epidermal growth factor receptor pathway substrate 8-related protein 3|EPS8R3Epidermal growth factor receptor kinase substrate 8-like protein 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TE67
- Q8TE67
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Epidermal growth factor receptor kinase substrate 8-like protein 3 protein (367-521AA)
- Recombinant Human Epidermal growth factor receptor kinase substrate 8-like protein 3 protein (367-521AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together