Anti-TBC1D15 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TBC1D15
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-766
- 77719-767
- Antigen Symbol:
- TBC1D15
- TBC1D15
- Antigen Synonyms:
- DKFZp686M1379|GAP for RAB7|Rab7-GAP|GTPase-activating protein RAB7|RAB7 GAP|Tre 2 budding uninhibited by benzimidazole cell division cycle 16 domain|TBC1 domain family member 15|FLJ12085|TBC15_HUMAN|FLJ33191|domain family member 15|Tre2/Bub2/Cdc16 domain family member 15|Tbc1d15|DKFZp761D0223
- DKFZp686M1379|GAP for RAB7|Rab7-GAP|GTPase-activating protein RAB7|RAB7 GAP|Tre 2 budding uninhibited by benzimidazole cell division cycle 16 domain|TBC1 domain family member 15|FLJ12085|TBC15_HUMAN|FLJ33191|domain family member 15|Tre2/Bub2/Cdc16 domain family member 15|Tbc1d15|DKFZp761D0223
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TC07
- Q8TC07
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human TBC1 domain family member 15 protein (283-404AA)
- Recombinant Human TBC1 domain family member 15 protein (283-404AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together