Anti-DBF4B Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: DBF4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77690-174
- 77719-706
- Antigen Symbol:
- DBF4B
- DBF4B
- Antigen Synonyms:
- FLJ13087|ZDBF1B|DBF 4B|chifb|ASK like protein 1|Dbf4-related factor 1|ASKL1|DBF4B_HUMAN|DBF4 homolog B|Dbf4 related factor 1|Chiffon homolog B|ASK-like protein 1|MGC15009|dbf4b|Activator of S phase kinase-like protein 1|DBF4 homolog B (S. cerevisiae)|Zinc finger DBF type containing 1B|Protein DBF4 homolog B|DRF1|Activator of S phase kinase like protein 1
- FLJ13087|ZDBF1B|DBF 4B|chifb|ASK like protein 1|Dbf4-related factor 1|ASKL1|DBF4B_HUMAN|DBF4 homolog B|Dbf4 related factor 1|Chiffon homolog B|ASK-like protein 1|MGC15009|dbf4b|Activator of S phase kinase-like protein 1|DBF4 homolog B (S. cerevisiae)|Zinc finger DBF type containing 1B|Protein DBF4 homolog B|DRF1|Activator of S phase kinase like protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NFT6
- Q8NFT6
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein DBF4 homolog B protein (1-300AA)
- Recombinant Human Protein DBF4 homolog B protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together