Anti-NSUN7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NSUN7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-671
- 77719-672
- Antigen Symbol:
- NSUN7
- NSUN7
- Antigen Synonyms:
- EC 2.1.1.-|NSUN7Putative methyltransferase NSUN7|NOL1/NOP2/Sun domain family member 7
- EC 2.1.1.-|NSUN7Putative methyltransferase NSUN7|NOL1/NOP2/Sun domain family member 7
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NE18
- Q8NE18
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Putative methyltransferase NSUN7 protein (1-300AA)
- Recombinant Human Putative methyltransferase NSUN7 protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together