Anti-KDM1B Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: KDM1B
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77719-631
- 77719-632
- Antigen Symbol:
- KDM1B
- KDM1B
- Antigen Synonyms:
- lysine (K)-specific demethylase 1B|FLJ43328|C6orf193|KDM1B|dJ298J15.2|Lysine-specific histone demethylase 1B|bA204B7.3|LSD2|FLJ34109|Lysine-specific histone demethylase 2|Lysine-specific demethylase 2|AOF1|amine oxidase (flavin containing) domain 1|Flavin-containing amine oxidase domain-containing protein 1|KDM1B_HUMAN|FLJ33898|amine oxidase, flavin containing 1
- lysine (K)-specific demethylase 1B|FLJ43328|C6orf193|KDM1B|dJ298J15.2|Lysine-specific histone demethylase 1B|bA204B7.3|LSD2|FLJ34109|Lysine-specific histone demethylase 2|Lysine-specific demethylase 2|AOF1|amine oxidase (flavin containing) domain 1|Flavin-containing amine oxidase domain-containing protein 1|KDM1B_HUMAN|FLJ33898|amine oxidase, flavin containing 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NB78
- Q8NB78
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lysine-specific histone demethylase 1B protein (451-544AA)
- Recombinant Human Lysine-specific histone demethylase 1B protein (451-544AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together