Order Entry
United States
ContactUsLinkComponent
Anti-PPM1K Rabbit Polyclonal Antibody (FITC)
Anti-PPM1K Rabbit Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-PPM1K Rabbit Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77719-501
  • 77719-500
  • Antigen Symbol:
  • PPM1K
  • PPM1K
  • Antigen Synonyms:
  • PP2C-kappa|PPM1K|BDP|PP2C-type mitochondrial phosphoprotein phosphatase|PP2C kappa|MSUDMV|Protein phosphatase Mg2+/Mn2+ dependent 1K|PP2C domain containing protein phosphatase 1K|PP2Ckappa|Protein phosphatase 1K|PP2C like mitochondrial protein|Protein phosphatase 2C isoform kappa|mitochondrial|PTMP|PP2CM|PPM1K_HUMAN|PP2C type mitochondrial phosphoprotein phosphatase|PP2C domain-containing protein phosphatase 1K|PP2C-like mitochondrial protein|UG0882E07
  • PP2C-kappa|PPM1K|BDP|PP2C-type mitochondrial phosphoprotein phosphatase|PP2C kappa|MSUDMV|Protein phosphatase Mg2+/Mn2+ dependent 1K|PP2C domain containing protein phosphatase 1K|PP2Ckappa|Protein phosphatase 1K|PP2C like mitochondrial protein|Protein phosphatase 2C isoform kappa|mitochondrial|PTMP|PP2CM|PPM1K_HUMAN|PP2C type mitochondrial phosphoprotein phosphatase|PP2C domain-containing protein phosphatase 1K|PP2C-like mitochondrial protein|UG0882E07
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q8N3J5
  • Q8N3J5
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Protein phosphatase 1K, mitochondrial protein (30-372AA)
  • Recombinant Human Protein phosphatase 1K, mitochondrial protein (30-372AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together