Anti-PLIN4 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLIN4
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-346
- 77719-347
- Antigen Symbol:
- PLIN4
- PLIN4
- Antigen Synonyms:
- Adipocyte protein S3-12|Plin 4|Plasma membrane associated protein S3 12|KIAA1881|PLIN4_HUMAN|PLIN4|Adipocyte protein S3 12|Perilipin-4|S3 12|hCG1646516
- Adipocyte protein S3-12|Plin 4|Plasma membrane associated protein S3 12|KIAA1881|PLIN4_HUMAN|PLIN4|Adipocyte protein S3 12|Perilipin-4|S3 12|hCG1646516
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96Q06
- Q96Q06
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Perilipin-4 protein (308-418AA)
- Recombinant Human Perilipin-4 protein (308-418AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together