Anti-MED30 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MED30
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-185
- 77719-184
- Antigen Symbol:
- MED30
- MED30
- Antigen Synonyms:
- THRAP6|TRAP/Mediator complex component TRAP25|MED30|MED30_HUMAN|Mediator complex subunit 30|Thyroid hormone receptor-associated protein 6|Trap25|Mediator of RNA polymerase II transcription subunit 30|Thyroid hormone receptor-associated protein complex 25 kDa component
- THRAP6|TRAP/Mediator complex component TRAP25|MED30|MED30_HUMAN|Mediator complex subunit 30|Thyroid hormone receptor-associated protein 6|Trap25|Mediator of RNA polymerase II transcription subunit 30|Thyroid hormone receptor-associated protein complex 25 kDa component
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96HR3
- Q96HR3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mediator of RNA polymerase II transcription subunit 30 protein (67-165AA)
- Recombinant Human Mediator of RNA polymerase II transcription subunit 30 protein (67-165AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together