Anti-PYCR2 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PYCR2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-064
- 77719-063
- Antigen Symbol:
- PYCR2
- PYCR2
- Antigen Synonyms:
- Pyrroline-5-carboxylate reductase 2|1810018M05Rik|Pyrroline 5 carboxylate reductase family member 2|Leftb|Pyrroline 5 carboxylate reductase 2|P5C reductase 2|PYCR2|PYCR 2|Pyrroline 5 carboxylate reductase isoform|OTTHUMP00000035614|FLJ54750|P5CR2|RP4-559A3.4|P5CR 2|P5CR2_HUMAN
- Pyrroline-5-carboxylate reductase 2|1810018M05Rik|Pyrroline 5 carboxylate reductase family member 2|Leftb|Pyrroline 5 carboxylate reductase 2|P5C reductase 2|PYCR2|PYCR 2|Pyrroline 5 carboxylate reductase isoform|OTTHUMP00000035614|FLJ54750|P5CR2|RP4-559A3.4|P5CR 2|P5CR2_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96C36
- Q96C36
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Pyrroline-5-carboxylate reductase 2 protein (2-320AA)
- Recombinant Human Pyrroline-5-carboxylate reductase 2 protein (2-320AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together