Anti-TP53INP1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: TP53INP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-011
- 77719-012
- Antigen Symbol:
- TP53INP1
- TP53INP1
- Antigen Synonyms:
- p53-dependent damage-inducible nuclear protein 1|Teap|TP53 DINP1|Tumor protein p53-inducible nuclear protein 1|p53 dependent damage inducible nuclear protein 1|FLJ22139|DKFZp434M1317|Stress induced protein|TP53INP1B|p53 inducible p53DINP1|SIP|p53 inducible nuclear protein 1|TP53INP1A|TP53DINP1|TP53INP1|Stress-induced protein|p53DINP1|TP53 INP1|T53I1_HUMAN|Tumor protein p53 inducible nuclear protein 1|p53 damage inducible nuclear protein 1
- p53-dependent damage-inducible nuclear protein 1|Teap|TP53 DINP1|Tumor protein p53-inducible nuclear protein 1|p53 dependent damage inducible nuclear protein 1|FLJ22139|DKFZp434M1317|Stress induced protein|TP53INP1B|p53 inducible p53DINP1|SIP|p53 inducible nuclear protein 1|TP53INP1A|TP53DINP1|TP53INP1|Stress-induced protein|p53DINP1|TP53 INP1|T53I1_HUMAN|Tumor protein p53 inducible nuclear protein 1|p53 damage inducible nuclear protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96A56
- Q96A56
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Tumor protein p53-inducible nuclear protein 1 protein (138-205AA)
- Recombinant Human Tumor protein p53-inducible nuclear protein 1 protein (138-205AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together