Anti-CAT1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CAT1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
Specifications
- Catalog No:
- 77718-981
- 77718-980
- Antigen Symbol:
- CAT1
- CAT1
- Antigen Synonyms:
- F2D10.11|At1g20630|F5M15.31Catalase-1|EC 1.11.1.6|CAT1
- F2D10.11|At1g20630|F5M15.31Catalase-1|EC 1.11.1.6|CAT1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- A. thaliana
- A. thaliana
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96528
- Q96528
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Arabidopsis thaliana Catalase-1 protein (1-492AA)
- Recombinant Arabidopsis thaliana Catalase-1 protein (1-492AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together