Anti-SORL1 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SORL1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77718-913
- 77718-914
- Antigen Symbol:
- SORL1
- SORL1
- Antigen Synonyms:
- Sortilin related receptor L(DLR class) A repeats containing|LR 11|C11orf32|SorLA 1|SorLA|SORL_HUMAN|LDLR relative with 11 ligand binding repeats|Mosaic protein LR11|Sortilin-related receptor|LDLR relative with 11 ligand-binding repeats|Sortilin related receptor|LRP9|Sorting protein-related receptor containing LDLR class A repeats|SORL 1|Low-density lipoprotein receptor relative with 11 ligand-binding repeats|SorLA-1|gp250|FLJ21930|FLJ39258|Sorting protein related receptor containing LDLR class A repeats|Low density lipoprotein receptor relative with 11 ligand binding repeats|LRP 9|LR11|SORL1
- Sortilin related receptor L(DLR class) A repeats containing|LR 11|C11orf32|SorLA 1|SorLA|SORL_HUMAN|LDLR relative with 11 ligand binding repeats|Mosaic protein LR11|Sortilin-related receptor|LDLR relative with 11 ligand-binding repeats|Sortilin related receptor|LRP9|Sorting protein-related receptor containing LDLR class A repeats|SORL 1|Low-density lipoprotein receptor relative with 11 ligand-binding repeats|SorLA-1|gp250|FLJ21930|FLJ39258|Sorting protein related receptor containing LDLR class A repeats|Low density lipoprotein receptor relative with 11 ligand binding repeats|LRP 9|LR11|SORL1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92673
- Q92673
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sortilin-related receptor protein (525-744AA)
- Recombinant Human Sortilin-related receptor protein (525-744AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together