Anti-RAPGEF5 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAPGEF5
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-896
- 77718-897
- Antigen Symbol:
- RAPGEF5
- RAPGEF5
- Antigen Synonyms:
- KIAA0277|MRas-regulated guanine nucleotide exchange factor|Guanine nucleotide exchange factor for Rap1|Related to Epac|GFR|M Ras regulated Rap GEF|MRGEF|M-Ras-regulated Rap GEF|Repac|RPGF5_HUMAN|Rapgef5|M-Ras-regulated GEF|Rap guanine nucleotide exchange factor 5|MR GEF|Rap guanine nucleotide exchange factor (GEF) 5|MR-GEF
- KIAA0277|MRas-regulated guanine nucleotide exchange factor|Guanine nucleotide exchange factor for Rap1|Related to Epac|GFR|M Ras regulated Rap GEF|MRGEF|M-Ras-regulated Rap GEF|Repac|RPGF5_HUMAN|Rapgef5|M-Ras-regulated GEF|Rap guanine nucleotide exchange factor 5|MR GEF|Rap guanine nucleotide exchange factor (GEF) 5|MR-GEF
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92565
- Q92565
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Rap guanine nucleotide exchange factor 5 protein (201-444AA)
- Recombinant Human Rap guanine nucleotide exchange factor 5 protein (201-444AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together