Anti-NR2E1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NR2E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77718-859
- 77718-860
- Antigen Symbol:
- NR2E1
- NR2E1
- Antigen Synonyms:
- Tailless homolog|NR2E1_HUMAN|NR2E1 / Tailless|hTll|Orphan nuclear receptor NR2E1|Protein tailless homolog|nr2e1|Nuclear receptor TLX|XTLL|TLX|Tll|Nuclear receptor subfamily 2 group E member 1
- Tailless homolog|NR2E1_HUMAN|NR2E1 / Tailless|hTll|Orphan nuclear receptor NR2E1|Protein tailless homolog|nr2e1|Nuclear receptor TLX|XTLL|TLX|Tll|Nuclear receptor subfamily 2 group E member 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y466
- Q9Y466
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Nuclear receptor subfamily 2 group E member 1 protein (106-385AA)
- Recombinant Human Nuclear receptor subfamily 2 group E member 1 protein (106-385AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together