Anti-EEF1AKMT1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EEF1AKMT1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-800
- 77718-799
- Antigen Symbol:
- EEF1AKMT1
- EEF1AKMT1
- Antigen Synonyms:
- EC 2.1.1.-|N6AMT2EEF1A lysine methyltransferase 1|Protein-lysine N-methyltransferase N6AMT2|EEF1AKMT1|N(6)-adenine-specific DNA methyltransferase 2
- EC 2.1.1.-|N6AMT2EEF1A lysine methyltransferase 1|Protein-lysine N-methyltransferase N6AMT2|EEF1AKMT1|N(6)-adenine-specific DNA methyltransferase 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WVE0
- Q8WVE0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human EEF1A lysine methyltransferase 1 protein (2-214AA)
- Recombinant Human EEF1A lysine methyltransferase 1 protein (2-214AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together