Anti-SH3TC2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SH3TC2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-711
- 77718-712
- Antigen Symbol:
- SH3TC2
- SH3TC2
- Antigen Synonyms:
- PP12494|D430044G18Rik|FLJ13605|SH3TC2|CMT4C|SH3 domain and tetratricopeptide repeats-containing protein 2|MNMN|RGD1309038|S3TC2_HUMAN|KIAA1985
- PP12494|D430044G18Rik|FLJ13605|SH3TC2|CMT4C|SH3 domain and tetratricopeptide repeats-containing protein 2|MNMN|RGD1309038|S3TC2_HUMAN|KIAA1985
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TF17
- Q8TF17
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human SH3 domain and tetratricopeptide repeat-containing protein 2 protein (188-427AA)
- Recombinant Human SH3 domain and tetratricopeptide repeat-containing protein 2 protein (188-427AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together