Anti-Tp53inp2 Rabbit Polyclonal Antibody
Catalog # 77708-101
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Tp53inp2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µl
- Form:Liquid
- Gene ID:Q8CFU8
- Antigen Synonyms:Tumor protein p53-inducible nuclear protein 2|Trp53inp2|Dor|Diabetes and obesity-regulated protein|Tp53inp2
- Storage Buffer:PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human TP53INP2
- Purification:Antigen affinity purified
- Cat. No.:77708-101
- Packaging:Vial
Specifications
About this item
Type:
Antigen: Tp53inp2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse