Anti-MAP2K7 (phospho-Ser271) Rabbit Polyclonal Antibody
100 µl, Vial
Catalog # 77707-678
Supplier: AFG Bioscience
Supplier #: A44451-100UL
Anti-MAP2K7 (phospho-Ser271) Rabbit Polyclonal Antibody
Catalog # 77707-678
Supplier: AFG Bioscience
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
WB:1:500-1:1000, IHC:1:50-1:100
Type:
Antigen: MAP2K7 (phospho-Ser271)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Size:100 µl
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Storage Buffer:PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77707-678
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice
- Antigen Symbol:MAP2K7 (phospho-Ser271)
- Gene ID:O14733
- Immunogen:Peptide sequence around phosphorylation site of Serine271(V-D-S(p)-K-A) derived from Human MAP2K7
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:PRKMK-7|PRKMK 7|Mitogen Activated Protein Kinase kinase 7|PRKMK7|MKK-7|JNK kinase 2|JNK activating kinase 2|Sek-2|SAPK kinase 4|Sek2|MEK 7|SAPKK-4|Dual specificity mitogen activated protein kinase kinase 7|JNK-activating kinase 2|Dual specificity mitogen-activated protein kinase kinase 7|stress-activated protein kinase kinase 4|MAPK/ERK kinase 7|Jnkk-2|MAPKK 7|SAPKK4|Sek 2|MKK7|MAPKK7|MKK 7|SKK4|c-Jun N-terminal kinase kinase 2|MAP2K7|MAP kinase kinase 7|MAPKK-7|JNKK 2|Jnkk2|MP2K7_HUMAN
- ELISA:Yes
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



