Anti-ARNT Rabbit Polyclonal Antibody
Catalog # 77705-208
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ARNT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P27540
- Antigen Synonyms:HIF-1-beta|Hypoxia Inducible Factor 1|Drnt|HIF1beta|HIF 1 beta|Hypoxia-inducible factor 1-beta|nuclear translocator|Class E basic helix-loop-helix protein 2|bHLHe2|ARNT protein|arnT|Dioxin receptor|HIF1B|ARNT_HUMAN|Tango|Hypoxia inducible factor 1 beta|HIF 1beta|Dioxin receptor nuclear translocator|HIF1-beta|Aryl hydrocarbon receptor nuclear translocator
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant protein
- Purification:Affinity purified
- Cat. No.:77705-208
- Packaging:Vial
Specifications
About this item
WB:1:1000-2000, IHC:1:200-500
Type:
Antigen: ARNT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat