Anti-TRIM72 Rabbit Polyclonal Antibody
Catalog # 77705-201
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TRIM72
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q6ZMU5
- Antigen Synonyms:Mg53|Tripartite motif-containing protein 72|TRI72_HUMAN|Mitsugumin-53|trim72|Tripartite motif containing 72|LOC493829|Tripartite motif containing protein 72
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant protein
- Purification:Affinity purified
- Cat. No.:77705-201
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TRIM72
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat