Anti-Histone H3 (Di-methyl-Lys9) Rabbit Polyclonal Antibody
77705-178
New Product
- Antigen Symbol:Histone H3 (Di-methyl-Lys9)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P68431/Q71DI3/P84243
- Antigen Synonyms:H3FA|HIST1H3J|H3FJ|HIST1H3E|HIST1H3G|HIST1H3B|Histone H3/f|H3FI|HIST1H3D|HIST1H3C|HIST1H3I|HIST1H3F|Histone H3/b|Histone H3/d|Histone H3.1|H3FH|H3FD|Histone H3/a|H3FF|Histone H3/|HIST1H3H|H3FC|H3FB|H3FL|Histone H3/c|H3FK|HIST1H3A
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthetic peptide
- Purification:Affinity purified
- Cat. No.:77705-178
- Packaging:Vial
WB:1:500-1:1000, ELISA:1:20000
Type:
Antigen: Histone H3 (Di-methyl-Lys9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat