Anti-TRAF4 Rabbit Polyclonal Antibody
Catalog # 77705-098
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TRAF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q9BUZ4
- Antigen Synonyms:Cysteine rich domain associated with RING and Traf domains protein 1|RNF 83|TRAF 4|TRAF4_HUMAN|cysteine rich domain associated with ring and TRAF domain|CART 1|Malignant 62|Metastatic lymph node gene 62 protein|Traf4|MLN 62|CART1|TNF receptor associated factor 4|RNF83|RING finger protein 83|tumor necrosis receptor-associated factor 4A|MLN62|Cysteine-rich domain associated with RING and Traf domains protein 1|TNF receptor-associated factor 4
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human TRAF4
- Purification:Antigen affinity purified
- Cat. No.:77705-098
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TRAF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat