Anti-CD40 (phospho-T254) Rabbit Polyclonal Antibody
Catalog # 77705-089
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CD40 (phospho-T254)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P25942
- Antigen Synonyms:TNF receptor superfamily member 5|TRAP|AI326936|p50|Tumor necrosis factor receptor superfamily member 5|IMD3|B-cell surface antigen CD40|CD40L receptor|Nerve growth factor receptor related B lymphocyte activation molecule|GP39|TNR5_HUMAN|OTTHUMP00000031700|CD40|CD40 molecule, TNF receptor superfamily member 5|IGM|MGC9013|T-BAM|CD40 protein|B cell-associated molecule|B cell surface antigen CD40|B cell associated molecule CD40|Bp50|CD40 antigen|TBAM|OTTHUMP00000031699|CDw40|CD 40|Tumor necrosis factor receptor superfamily, member 5|Tumor necrosis factor receptor superfamily, member 5, isoform CRA_a|HIGM1|CD40 molecule|CD40 antigen (TNF receptor superfamily member 5)|CD40 type II isoform|Tumor necrosis factor receptor superfamily member 5 precursor|TNFRSF5
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human CD40 around the phosphorylation site of T254
- Purification:Antigen affinity purified
- Cat. No.:77705-089
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:5000
Type:
Antigen: CD40 (phospho-T254)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse