Anti-MED26 Rabbit Polyclonal Antibody
Catalog # 77705-067
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MED26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O95402
- Antigen Synonyms:Mediator of RNA polymerase II transcription subunit 26|ARC70|Cofactor required for Sp1 transcriptional activation, subunit 7, 70kDa|Activator recruited cofactor 70 kDa component|Med26|CRSP complex subunit 7|MED 26|CRSP70|ARC 70|Transcriptional coactivator CRSP70|MED26_HUMAN|Activator-recruited cofactor 70 kDa component|Mediator complex subunit 26|CRSP 7|Cofactor required for Sp1 transcriptional activation, subunit 7 (70kD)|Cofactor required for Sp1 transcriptional activation subunit 7
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human CRSP70
- Purification:Antigen affinity purified
- Cat. No.:77705-067
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: MED26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse