Anti-ZMYM4 Rabbit Polyclonal Antibody
Catalog # 77705-012
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ZMYM4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q5VZL5
- Antigen Synonyms:KIAA0425|Zinc finger MYM-type protein 4|ZMYM4|Zinc finger protein 262|ZNF262
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ZNF262
- Purification:Antigen affinity purified
- Cat. No.:77705-012
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: ZMYM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse