Anti-RPS19BP1 Rabbit Polyclonal Antibody
Catalog # 77704-997
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RPS19BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q86WX3
- Antigen Synonyms:RPS19BP1|Active regulator of SIRT1|AROS_HUMAN|Ribosomal protein S19 binding protein 1|S19BP|40S ribosomal protein S19-binding protein 1|40S ribosomal protein S19 binding protein 1|RPS19 binding protein 1|RPS19-binding protein 1|AROS|Homolog of mouse S19 binding protein
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Ribosomal Protein S19BP
- Purification:Antigen affinity purified
- Cat. No.:77704-997
- Packaging:Vial
Specifications
About this item
Type:
Antigen: RPS19BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human