Anti-UTP15 Rabbit Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: UTP15
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77722-196
- 77722-195
- Antigen Symbol:
- UTP15
- UTP15
- Antigen Synonyms:
- FLJ12787|UTP15, U3 small nucleolar ribonucleoprotein, homolog (yeast)|U3 small nucleolar RNA-associated protein 15 homolog|UTP15, U3 small nucleolar ribonucleoprotein, homolog (S. cerevisiae)|UTP15|Src-associated protein SAW|NET21|UTP15_HUMAN|U3 small nucleolar ribonucleoprotein
- FLJ12787|UTP15, U3 small nucleolar ribonucleoprotein, homolog (yeast)|U3 small nucleolar RNA-associated protein 15 homolog|UTP15, U3 small nucleolar ribonucleoprotein, homolog (S. cerevisiae)|UTP15|Src-associated protein SAW|NET21|UTP15_HUMAN|U3 small nucleolar ribonucleoprotein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TED0
- Q8TED0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human U3 small nucleolar RNA-associated protein 15 homolog protein (234-518AA)
- Recombinant Human U3 small nucleolar RNA-associated protein 15 homolog protein (234-518AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together