Anti-TAS1R2 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TAS1R2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-184
- 77722-185
- Antigen Symbol:
- TAS1R2
- TAS1R2
- Antigen Synonyms:
- G-protein coupled receptor 71|Sweet taste receptor T1R2|TAS1R2|TS1R2_HUMAN|Taste receptor type 1 member 2|T1R2|G protein coupled receptor 71|TR2|GPR71
- G-protein coupled receptor 71|Sweet taste receptor T1R2|TAS1R2|TS1R2_HUMAN|Taste receptor type 1 member 2|T1R2|G protein coupled receptor 71|TR2|GPR71
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TE23
- Q8TE23
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Taste receptor type 1 member 2 protein (183-362AA)
- Recombinant Human Taste receptor type 1 member 2 protein (183-362AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together