Order Entry
United States
ContactUsLinkComponent
Anti-UHMK1 Rabbit Polyclonal Antibody
Anti-UHMK1 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-UHMK1 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77690-280
  • 77722-129
  • Antigen Symbol:
  • UHMK1
  • UHMK1
  • Antigen Synonyms:
  • UHMK1_HUMAN|P-CIP2|U2AF homology motif kinase 1|Kinase interacting with stathmin|kinase interacting with leukemia-associated gene (stathmin)|KIS protein kinase|FLJ23015|KIS|OTTHUMP00000029529|UHMK1|U2AF homology motif (UHM) kinase 1|Serine/threonine-protein kinase Kist|KIST|Serine threonine protein kinase Kist|DKFZp434C1613
  • UHMK1_HUMAN|P-CIP2|U2AF homology motif kinase 1|Kinase interacting with stathmin|kinase interacting with leukemia-associated gene (stathmin)|KIS protein kinase|FLJ23015|KIS|OTTHUMP00000029529|UHMK1|U2AF homology motif (UHM) kinase 1|Serine/threonine-protein kinase Kist|KIST|Serine threonine protein kinase Kist|DKFZp434C1613
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q8TAS1
  • Q8TAS1
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Serine/threonine-protein kinase Kist protein (183-385AA)
  • Recombinant Human Serine/threonine-protein kinase Kist protein (183-385AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together