Anti-NSUN5 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NSUN5
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-835
- 77721-836
- Antigen Symbol:
- NSUN5
- NSUN5
- Antigen Synonyms:
- NOP2/Sun domain family, member 5A|NSUN5|Putative methyltransferase NSUN5|NSUN5_HUMAN|NOL1|Williams-Beuren syndrome chromosomal region 20A protein|NOL1R|NOL1/NOP2/Sun domain family member 5|MGC986|WBSCR20A|p120|WBSCR20|FLJ10267|Williams Beuren syndrome chromosome region 20A|NOP2/Sun domain family, member 5|Williams-Beuren syndrome critical region protein 20 copy A|NOL1-related protein
- NOP2/Sun domain family, member 5A|NSUN5|Putative methyltransferase NSUN5|NSUN5_HUMAN|NOL1|Williams-Beuren syndrome chromosomal region 20A protein|NOL1R|NOL1/NOP2/Sun domain family member 5|MGC986|WBSCR20A|p120|WBSCR20|FLJ10267|Williams Beuren syndrome chromosome region 20A|NOP2/Sun domain family, member 5|Williams-Beuren syndrome critical region protein 20 copy A|NOL1-related protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96P11
- Q96P11
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Probable 28S rRNA (cytosine-C(5))-methyltransferase protein (362-466AA)
- Recombinant Human Probable 28S rRNA (cytosine-C(5))-methyltransferase protein (362-466AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together