Anti-COG3 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: COG3
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-777
- 77721-778
- Antigen Symbol:
- COG3
- COG3
- Antigen Synonyms:
- Cog3|p94|Conserved oligomeric Golgi complex subunit 3|SEC34, S. cerevisiae, homolog of|COG3_HUMAN|Component of oligomeric Golgi complex 3|SEC34|Vesicle-docking protein SEC34 homolog|Tethering factor SEC34|Vesicle docking protein SEC34 homolog|OTTHUMP00000018352|Component of golgi transport complex 3|COG complex subunit 3|SEC34 S. cerevisiae homolog of
- Cog3|p94|Conserved oligomeric Golgi complex subunit 3|SEC34, S. cerevisiae, homolog of|COG3_HUMAN|Component of oligomeric Golgi complex 3|SEC34|Vesicle-docking protein SEC34 homolog|Tethering factor SEC34|Vesicle docking protein SEC34 homolog|OTTHUMP00000018352|Component of golgi transport complex 3|COG complex subunit 3|SEC34 S. cerevisiae homolog of
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96JB2
- Q96JB2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Conserved oligomeric Golgi complex subunit 3 protein (101-400AA)
- Recombinant Human Conserved oligomeric Golgi complex subunit 3 protein (101-400AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together