Anti-DCUN1D1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DCUN1D1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-708
- 77721-709
- Antigen Symbol:
- DCUN1D1
- DCUN1D1
- Antigen Synonyms:
- DCN1|Defective in cullin neddylation protein 1-like protein 1|DCUN1D1|DCUN1 domain-containing protein 1|RP42|Squamous cell carcinoma-related oncogene|DCUN1L1|SCCRODCN1-like protein 1
- DCN1|Defective in cullin neddylation protein 1-like protein 1|DCUN1D1|DCUN1 domain-containing protein 1|RP42|Squamous cell carcinoma-related oncogene|DCUN1L1|SCCRODCN1-like protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96GG9
- Q96GG9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DCN1-like protein 1 protein (1-259AA)
- Recombinant Human DCN1-like protein 1 protein (1-259AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together