Anti-LINGO1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: LINGO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77721-686
- 77721-687
- Antigen Symbol:
- LINGO1
- LINGO1
- Antigen Synonyms:
- Lrrn 6a|LRR and Ig domain containing Nogo Receptor interating protein|Leucine rich repeat neuronal protein 6A|FLJ14594|Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 1|Leucine rich repeat neuronal 6A|LERN1|LINGO1|Lrrn6a|Leucine-rich repeat and immunoglobilin domain-containing protein 1|LIGO1_HUMAN|UNQ201|Leucine-rich repeat neuronal protein 1|PRO227|Leucine rich repeat neuronal protein 1|Leucine rich repeat and Ig domain containing 1|Leucine-rich repeat neuronal protein 6A|MGC17422|Lingo 1|LERN 1|Lrrn6a protein|Nogo Receptor interacting protein
- Lrrn 6a|LRR and Ig domain containing Nogo Receptor interating protein|Leucine rich repeat neuronal protein 6A|FLJ14594|Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 1|Leucine rich repeat neuronal 6A|LERN1|LINGO1|Lrrn6a|Leucine-rich repeat and immunoglobilin domain-containing protein 1|LIGO1_HUMAN|UNQ201|Leucine-rich repeat neuronal protein 1|PRO227|Leucine rich repeat neuronal protein 1|Leucine rich repeat and Ig domain containing 1|Leucine-rich repeat neuronal protein 6A|MGC17422|Lingo 1|LERN 1|Lrrn6a protein|Nogo Receptor interacting protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96FE5
- Q96FE5
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 1 protein (42-240AA)
- Recombinant Human Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 1 protein (42-240AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together