Anti-COQ8B Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: COQ8B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77690-256
- 77721-654
- Antigen Symbol:
- COQ8B
- COQ8B
- Antigen Synonyms:
- NPHS9|ADCK4|COQ8B|Atypical kinase COQ8B mitochondrial|aarF domain containing protein kinase 4|Coenzyme Q8B|ADCK4_HUMAN|Uncharacterized aarF domain containing protein kinase 4|Uncharacterized aarF domain-containing protein kinase 4
- NPHS9|ADCK4|COQ8B|Atypical kinase COQ8B mitochondrial|aarF domain containing protein kinase 4|Coenzyme Q8B|ADCK4_HUMAN|Uncharacterized aarF domain containing protein kinase 4|Uncharacterized aarF domain-containing protein kinase 4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96D53
- Q96D53
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Atypical kinase COQ8B, mitochondrial protein (10-137AA)
- Recombinant Human Atypical kinase COQ8B, mitochondrial protein (10-137AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together