Anti-SHC3 Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:500-1:5000
Type:
Antigen: SHC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77721-447
- 77721-446
- Antigen Symbol:
- SHC3
- SHC3
- Antigen Synonyms:
- SHCC|Src homology 2 domain-containing-transforming protein C3|N-Shc|SH2 domain protein C3|SHC adaptor protein 3|Protein Rai|Shc3|SHC protein C|Src homology 2 domain-containing transforming protein|SHC-like protein, neuronal|Rai|SHC3_HUMAN|SHC-transforming protein C|Shc3 p51|NSHC|Neuronal Shc|SHC (Src homology 2 domain containing) transforming protein 3|N SHC|SHC-transforming protein 3
- SHCC|Src homology 2 domain-containing-transforming protein C3|N-Shc|SH2 domain protein C3|SHC adaptor protein 3|Protein Rai|Shc3|SHC protein C|Src homology 2 domain-containing transforming protein|SHC-like protein, neuronal|Rai|SHC3_HUMAN|SHC-transforming protein C|Shc3 p51|NSHC|Neuronal Shc|SHC (Src homology 2 domain containing) transforming protein 3|N SHC|SHC-transforming protein 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92529
- Q92529
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human SHC-transforming protein 3 protein (323-468AA)
- Recombinant Human SHC-transforming protein 3 protein (323-468AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together